missing translation for 'onlineSavingsMsg'
Learn More

4'-(Methylthio)acetophenone 98.0+%, TCI America™

Catalog No. M16725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16725G 5 g
M167225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16725G Supplier TCI America Supplier No. M16725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1778-09-2
Molecular Formula C9H10OS
Molecular Weight (g/mol) 166.24
MDL Number MFCD00039835
InChI Key JECUZQLBQKNEMW-UHFFFAOYSA-N
Synonym 4'-methylthio acetophenone, 4-methylthio acetophenone, 4'-methylthioacetophenone, 1-4-methylsulfanyl phenyl ethanone, 4-acetyl thioanisole, 4'-methylmercapto acetophenone, 1-4-methylthio phenyl ethanone, 1-4-methylthio phenyl ethan-1-one, 1-4-methylsulfanylphenyl ethanone
PubChem CID 74501
IUPAC Name 1-[4-(methylsulfanyl)phenyl]ethan-1-one
SMILES CSC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 79°C
Boiling Point 135°C
Quantity 5 g
Formula Weight 166.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(Methylthio)acetophenone