CAS RN 1774-35-2
CAS RN 1774-35-2
IUPAC Name:
1-methyl-4-(4-methylbenzenesulfinyl)benzene
Synonyms:
1-methyl-4-(4-methylbenzenesulfinyl)benzene
Molecular Weight (g/mol):
230.33
Molecular Formula:
C14H14OS
InChi Key:
MJWNJEJCQHNDNM-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
p-Tolyl Sulfoxide 98.0+%, TCI America™
CAS: 1774-35-2 Molecular Formula: C14H14OS Molecular Weight (g/mol): 230.325 MDL Number: MFCD00008546 InChI Key: MJWNJEJCQHNDNM-UHFFFAOYSA-N Synonym: Di-p-tolyl Sulfoxide PubChem CID: 15680 IUPAC Name: 1-methyl-4-(4-methylphenyl)sulfinylbenzene SMILES: CC1=CC=C(C=C1)S(=O)C2=CC=C(C=C2)C