missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl Sulfoxide 98.0+%, TCI America™

Catalog No. T107525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T107525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T107525G Supplier TCI America Supplier No. T107525G

Chemical Identifiers

CAS 1774-35-2
Molecular Formula C14H14OS
Molecular Weight (g/mol) 230.325
MDL Number MFCD00008546
InChI Key MJWNJEJCQHNDNM-UHFFFAOYSA-N
Synonym Di-p-tolyl Sulfoxide
PubChem CID 15680
IUPAC Name 1-methyl-4-(4-methylphenyl)sulfinylbenzene
SMILES CC1=CC=C(C=C1)S(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 94°C
Color White
Quantity 25 g
Formula Weight 230.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Tolyl Sulfoxide
RTECSNumber XT4810000
TSCA No