accessibility menu, dialog, popup

UserName

CAS RN 177080-67-0

H3CH2COOCH3

CAS RN 177080-67-0

IUPAC Name: 2-methyladamantan-2-yl 2-methylprop-2-enoate
Synonyms: 2-methyladamantan-2-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: FDYDISGSYGFRJM-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC1(C)C2CC3CC(C2)CC1C3
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results