missing translation for 'onlineSavingsMsg'
Learn More

2-Methacryloyloxy-2-methyladamantane (stabilized with MEHQ) 97.0+%, TCI America™

Catalog No. M22605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22605G 5 g
M22601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22605G Supplier TCI America Supplier No. M22605G
Only null left

Chemical Identifiers

CAS 177080-67-0
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD04114354
InChI Key FDYDISGSYGFRJM-UHFFFAOYSA-N
Synonym 2-Methyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Methyl-2-adamantyl Ester
PubChem CID 9837566
IUPAC Name (2-methyl-2-adamantyl) 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1(C2CC3CC(C2)CC1C3)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 234.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methacryloyloxy-2-methyladamantane (stabilized with MEHQ)
EINECSNumber (4)-1884
TSCA No
Recommended Storage Freezer