accessibility menu, dialog, popup

UserName

CH3ClBOHHO

CAS RN 175883-63-3

IUPAC Name: (3-chloro-4-methylphenyl)boronic acid
Synonyms: 3-chloro-4-methylphenylboronic acid
Molecular Weight (g/mol): 170.4
Molecular Formula: C7H8BClO2
InChi Key: YTJUYWRCAZWVSX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1Cl)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)