missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C25251G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25251G Supplier TCI America Supplier No. C25251G

Specifications

Chemical Name or Material 3-Chloro-4-methylphenylboronic Acid (contains varying amounts of Anhydride)
Melting Point 247°C
CAS 175883-63-3
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C7H8BClO2
MDL Number MFCD04039010
Synonym 3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
InChI Key YTJUYWRCAZWVSX-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1Cl)B(O)O
IUPAC Name (3-chloro-4-methylphenyl)boronic acid
Molecular Weight (g/mol) 170.40
PubChem CID 3854610
Formula Weight 170.40
Show More Show Less