accessibility menu, dialog, popup

UserName

CAS RN 1758-25-4

IUPAC Name: 2-(2,5-dimethoxyphenyl)acetic acid
Synonyms: (2,5-dimethoxyphenyl)acetic acid
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: BBZDYQUXRFATHZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(CC(O)=O)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)