missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. D14455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14455G 5 g
D144525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14455G Supplier TCI America Supplier No. D14455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1758-25-4
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00004322
InChI Key BBZDYQUXRFATHZ-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenylacetic acid, 2-2,5-dimethoxyphenyl acetic acid, 2,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,5-dimethoxy, acetic acid, 2,5-dimethoxyphenyl, 2,5-dimethoxyphenyl-acetic acid, acmc-209eai, 2,5-dimethoxyphenyl, rarechem al bo 0334, 2,5-dimethoxy-phenethyl acid
PubChem CID 74469
IUPAC Name 2-(2,5-dimethoxyphenyl)acetic acid
SMILES COC1=CC(=C(C=C1)OC)CC(=O)O

Specifications

Melting Point 124°C
Color Yellow
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethoxyphenylacetic Acid