accessibility menu, dialog, popup

UserName

CAS RN 1750-12-5

IUPAC Name: 4-amino-3-hydrazinyl-4,5-dihydro-1H-1,2,4-triazole-5-thione
Synonyms: 4-amino-5-hydrazinyl-2H-1,2,4-triazole-3-thione
Molecular Weight (g/mol): 146.17
Molecular Formula: C2H6N6S
InChi Key: RDIMQHBOTMWMJA-UHFFFAOYSA-N
SMILES: NNC1=NNC(=S)N1N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)