missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole 95.0+%, TCI America™

Catalog No. A09565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09565G 5 g
A095625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09565G Supplier TCI America Supplier No. A09565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1750-12-5
Molecular Formula C2H6N6S
Molecular Weight (g/mol) 146.172
MDL Number MFCD00003098
InChI Key RDIMQHBOTMWMJA-UHFFFAOYSA-N
Synonym 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiol
PubChem CID 2723946
IUPAC Name 4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione
SMILES C1(=S)NN=C(N1N)NN

Specifications

Melting Point 240°C
Color White
Quantity 5 g
UN Number 1325
Formula Weight 146.17
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole [for Determination of Aldehydes]
EINECSNumber (9)-1091
TSCA Yes