accessibility menu, dialog, popup

UserName

CAS RN 17407-09-9

IUPAC Name: 2-[(trimethylsilyl)oxy]ethyl 2-methylprop-2-enoate
Synonyms: 2-[(trimethylsilyl)oxy]ethyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 202.33
Molecular Formula: C9H18O3Si
InChi Key: WUGOQZFPNUYUOO-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCO[Si](C)(C)C
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)