accessibility menu, dialog, popup

UserName

CAS RN 1738-36-9

IUPAC Name: 2-methoxyacetonitrile
Synonyms: acetonitrile, methoxy-
Molecular Weight (g/mol): 71.08
Molecular Formula: C3H5NO
InChi Key: QKPVEISEHYYHRH-UHFFFAOYSA-N
SMILES: COCC#N
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)