missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetonitrile 98.0+%, TCI America™

Catalog No. M01035ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M01035ML 5 mL
M010325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M01035ML Supplier TCI America Supplier No. M01035ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1738-36-9
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00001892
InChI Key QKPVEISEHYYHRH-UHFFFAOYSA-N
PubChem CID 74442
IUPAC Name 2-methoxyacetonitrile
SMILES COCC#N

Specifications

Color Yellow
Boiling Point 119°C
Quantity 5 mL
UN Number 1993
Synonym methoxyacetonitrile, acetonitrile, methoxy, acetonitrile, 2-methoxy, ncch2och3, 2-methoxy acetonitrile, 2-methoxy-acetonitrile, acetonitrile,2-methoxy, intermediates-zcf02294, acmc-209e7f
Formula Weight 71.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methoxyacetonitrile