CAS RN 172975-69-8
IUPAC Name:
(3,5-dimethylphenyl)boronic acid
Synonyms:
3,5-dimethylphenylboronic acid
Molecular Weight (g/mol):
149.98
Molecular Formula:
C8H11BO2
InChi Key:
DJGHSJBYKIQHIK-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC(C)=C1)B(O)O
1
–
2
of
2
results
1
Promotions Available
3,5-Dimethylbenzeneboronic acid, 98%
3,5-Dimethylbenzeneboronic acid, 98%
CAS: 172975-69-8 Molecular Formula: C8H11BO2 Molecular Weight (g/mol): 149.984 MDL Number: MFCD00185689 InChI Key: DJGHSJBYKIQHIK-UHFFFAOYSA-N Synonym: 3,5-dimethylphenyl boronic acid,3,5-dimethylbenzeneboronic acid,3,5-dimethylphenyl boranediol,m-xylene-5-boronic acid,boronic acid, 3,5-dimethylphenyl,pubchem1833,acmc-209e5l,ksc174i6l,3.5-dimethylphenylboronic acid PubChem CID: 2734349 IUPAC Name: (3,5-dimethylphenyl)boronic acid SMILES: B(C1=CC(=CC(=C1)C)C)(O)O
2
3,5-Dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
CAS: 172975-69-8 | C8H11BO2 | 149.984 g/mol