missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzeneboronic acid, 98%

Catalog No. AAB2343414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2343414 25 g
AAB2343403 1 g
AAB2343406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2343414 Supplier Thermo Scientific Chemicals Supplier No. B2343414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 172975-69-8 | C8H11BO2 | 149.984 g/mol

3,5-Dimethylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Chemical Identifiers

CAS 172975-69-8
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.984
MDL Number MFCD00185689
InChI Key DJGHSJBYKIQHIK-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl boronic acid, 3,5-dimethylbenzeneboronic acid, 3,5-dimethylphenyl boranediol, m-xylene-5-boronic acid, boronic acid, 3,5-dimethylphenyl, pubchem1833, acmc-209e5l, ksc174i6l, 3.5-dimethylphenylboronic acid
PubChem CID 2734349
IUPAC Name (3,5-dimethylphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)C)C)(O)O

Specifications

Melting Point 235°C to 237°C
Quantity 25 g
Beilstein 7368658
Formula Weight 149.99
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only