CAS RN 1719-19-3
IUPAC Name:
2-methyl-4-phenylbut-3-yn-2-ol
Synonyms:
2-methyl-4-phenylbut-3-yn-2-ol
Molecular Weight (g/mol):
160.22
Molecular Formula:
C11H12O
InChi Key:
FUPXYICBZMASCM-UHFFFAOYSA-N
SMILES:
CC(C)(O)C#CC1=CC=CC=C1
1
–
1
of
1
results
1
2-Methyl-4-phenyl-3-butyn-2-ol 98.0+%, TCI America™
CAS: 1719-19-3 Molecular Formula: C11H12O Molecular Weight (g/mol): 160.22 MDL Number: MFCD00041572 InChI Key: FUPXYICBZMASCM-UHFFFAOYSA-N Synonym: 2-Hydroxy-2-methyl-4-phenyl-3-butyne PubChem CID: 288327 IUPAC Name: 2-methyl-4-phenylbut-3-yn-2-ol SMILES: CC(C)(O)C#CC1=CC=CC=C1