missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4-phenyl-3-butyn-2-ol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 54°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 160.22 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Methyl-4-phenyl-3-butyn-2-ol |
Safety and Handling
| TSCA | No |