accessibility menu, dialog, popup

UserName

CAS RN 171860-68-7

IUPAC Name: bromozincylium; cyclopentanide
Synonyms: bromozincylium; cyclopentanide
Molecular Weight (g/mol): 214.41
Molecular Formula: C5H9BrZn
InChi Key: GTJUPSNUGOBNMF-UHFFFAOYSA-M
SMILES: [Zn+]Br.C1CC[CH-]C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results