missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58764AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58764AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58764AD Supplier Thermo Scientific Chemicals Supplier No. H58764AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 171860-68-7 | C5H9BrZn | 214.411 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active nucleophile in Pd-catalyzed cross-coupling of heteroaryl halides.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and

Chemical Identifiers

CAS 171860-68-7
Molecular Formula C5H9BrZn
Molecular Weight (g/mol) 214.411
MDL Number MFCD00671991
InChI Key GTJUPSNUGOBNMF-UHFFFAOYSA-M
Synonym zinc, bromocyclopentyl, cyclopentylzinc bromide, cyclopentylzinc bromide solution, bromo cyclopentyl zinc, bromo cyclopentylzinc, zinc,bromocyclopentyl, cyclopentyl zinc bromide, cyclopentylzinc ii bromide, gopycbpmccjysn-uhfffaoysa-m
PubChem CID 4139106
IUPAC Name bromozinc(1+);cyclopentane
SMILES C1CC[CH-]C1.[Zn+]Br

Specifications

Density 0.955
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Reacts with water.
Formula Weight 214.41
Chemical Name or Material Cyclopentylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only