accessibility menu, dialog, popup

UserName

CH3(R)O(S)ONNOHNHN(R)FFFFFFF

CAS RN 170729-80-3

IUPAC Name: 5-{[(2S,3R)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl}-2,3-dihydro-1H-1,2,4-triazol-3-one
Synonyms: aprepitant
Molecular Weight (g/mol): 534.44
Molecular Formula: C23H21F7N4O3
InChi Key: ATALOFNDEOCMKK-BYYRLHKVSA-N
SMILES: C[C@@H](O[C@@H]1OCCN(CC2=NC(=O)NN2)[C@@H]1C1=CC=C(F)C=C1)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (2)
Product Type 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)