missing translation for 'onlineSavingsMsg'
Learn More

Aprepitant 98.0+%, TCI America™

Catalog No. A3135500MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3135500MG 500 mg
A3135100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3135500MG Supplier TCI America Supplier No. A3135500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 170729-80-3
Molecular Formula C23H21F7N4O3
Molecular Weight (g/mol) 534.435
MDL Number MFCD08277635
InChI Key ATALOFNDEOCMKK-OITMNORJSA-N
Synonym 5-[[(2R,3S)-2-[(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3-one
PubChem CID 6918365
ChEBI CHEBI:499361
IUPAC Name 5-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl]-1,2-dihydro-1,2,4-triazol-3-one
SMILES CC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)OC2C(N(CCO2)CC3=NC(=O)NN3)C4=CC=C(C=C4)F

Specifications

Melting Point 252°C
Color White
Quantity 500 mg
Formula Weight 534.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Aprepitant
RTECSNumber XZ5480400
TSCA No
Recommended Storage Refrigerator