missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, N6,O2'-Dibutyryl-, Sodium Salt

Catalog No. 28745100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28745100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28745100MG Supplier MilliporeSigma™ Supplier No. 28745100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Dibutyryl-cAMP, CAS 16980-89-5, is a cell-permeable cAMP analog that preferentially activates PKA.

Cell Permeable

Specifications

Color White
Target Proteins PKA
CAS 16980-89-5
Molecular Formula C18H23N5O8P·Na
Formula Weight 491.4g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 100 mg
Purity 95% by HPLC