CAS RN 16969-10-1
IUPAC Name:
propyl (2Z)-2-hydroxy-3-phenoxyprop-2-enoate
Synonyms:
propyl (2Z)-2-hydroxy-3-phenoxyprop-2-enoate
Molecular Weight (g/mol):
222.24
Molecular Formula:
C12H14O4
InChi Key:
FOKZHJCFBNVOAV-LUAWRHEFSA-N
SMILES:
CCCOC(=O)C(\O)=C\OC1=CC=CC=C1
1
–
1
of
1
results
1
2-Hydroxy-3-phenoxypropyl Acrylate (stabilized with MEHQ) 85.0+%, TCI America™
CAS: 16969-10-1 Molecular Formula: C12H14O4 Molecular Weight (g/mol): 222.24 MDL Number: MFCD00066300 InChI Key: FOKZHJCFBNVOAV-LUAWRHEFSA-N Synonym: Acrylic Acid 2-Hydroxy-3-phenoxypropyl Ester PubChem CID: 4443516 IUPAC Name: propyl (2Z)-2-hydroxy-3-phenoxyprop-2-enoate SMILES: CCCOC(=O)C(\O)=C\OC1=CC=CC=C1