missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-phenoxypropyl Acrylate (stabilized with MEHQ) 85.0+%, TCI America™

Catalog No. H1614500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1614500G 500 g
H161425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1614500G Supplier TCI America Supplier No. H1614500G
Only null left

Chemical Identifiers

CAS 16969-10-1
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00066300
InChI Key FOKZHJCFBNVOAV-LUAWRHEFSA-N
Synonym Acrylic Acid 2-Hydroxy-3-phenoxypropyl Ester
PubChem CID 4443516
IUPAC Name propyl (2Z)-2-hydroxy-3-phenoxyprop-2-enoate
SMILES CCCOC(=O)C(\O)=C\OC1=CC=CC=C1

Specifications

Color Yellow
Quantity 500 g
Formula Weight 222.24
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-3-phenoxypropyl Acrylate (stabilized with MEHQ)
EINECSNumber (3)-0565
TSCA Yes