missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium ICP Standard, 10,000 ppm Ti in H2O/tr HF, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 10,000ppm Ti |
| CAS Min % | 94.0, 3.89, 0.09 |
| CAS Max % | 98.0, 4.05, 0.1 |
| Equivalence | 1 mL = 10 mg Ti |
| Chemical Name or Material | Titanium ICP Standard |