missing translation for 'onlineSavingsMsg'
Learn More

Titanium ICP Standard, 1000 ppm Ti in H2O/tr HF, Ricca Chemical

Catalog No. p-3252585
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PTI1KW100 Natural LDPE Bottle
PTI1KW500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PTI1KW100 Supplier Ricca Chemical Company Supplier No. PTI1KW100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16962-40-6 , 7664-39-3 , 7732-18-5

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Ti
CAS Min % 98.0, 0.4, 0.01
CAS Max % 100.0, 0.42, 0.01
Equivalence 1 mL = 1 mg Ti
Chemical Name or Material Titanium ICP Standard