accessibility menu, dialog, popup

UserName

CAS RN 16921-30-5

IUPAC Name: dipotassium hexachloroplatinumdiuide
Synonyms: potassium hexachloroplatinate
Molecular Weight (g/mol): 485.98
Molecular Formula: Cl6K2Pt
InChi Key: DPAIVKJGTXERIM-UHFFFAOYSA-H
SMILES: [K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (9)
  • (1)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (3)
  • (2)
Grade 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (6)
  • (9)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)