missing translation for 'onlineSavingsMsg'
Learn More

Platinum AA Standard, 1000 ppm Pt in 3% HCl, Ricca Chemical

Catalog No. APT1KH100
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
APT1KH100 Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APT1KH100 Supplier Ricca Chemical Company Supplier No. APT1KH100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16921-30-5 , 7440-06-4 , 7732-18-5
Molecular Formula Cl6K2Pt
Molecular Weight (g/mol) 485.981
InChI Key DPAIVKJGTXERIM-UHFFFAOYSA-H
PubChem CID 61856
IUPAC Name dipotassium;hexachloroplatinum(2-)
SMILES Cl[Pt-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Pt
CAS Min % 93.0, 4.38, 0.09
CAS Max % 97.0, 4.56, 0.1
Equivalence 1 mL = 1 mg Pt
Chemical Name or Material Platinum AA Standard