accessibility menu, dialog, popup

UserName

OSON

CAS RN 16851-82-4

IUPAC Name: 1-(benzenesulfonyl)-1H-pyrrole
Synonyms: 1-(benzenesulfonyl)pyrrole
Molecular Weight (g/mol): 207.25
Molecular Formula: C10H9NO2S
InChi Key: PPPXRIUHKCOOMU-UHFFFAOYSA-N
SMILES: O=S(=O)(N1C=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)