missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Phenylsulfonyl)pyrrole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 89°C |
| Color | White |
| Quantity | 5 g |
| Synonym | 1-phenylsulfonyl pyrrole, 1-phenylsulfonyl-1h-pyrrole, n-benzenesulphonylpyrrole, 1-benzenesulfonyl pyrrole, n-benzenesulfonylpyrrole, 1-benzenesulfonyl-1h-pyrrole, n-phenylsulfonylpyrrole, phenylsulfonyl pyrrole, 1h-pyrrole,1-phenylsulfonyl, 1h-pyrrole, 1-phenylsulfonyl |
| Formula Weight | 207.25 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-(Phenylsulfonyl)pyrrole |
Safety and Handling
| TSCA | No |