accessibility menu, dialog, popup

UserName

CAS RN 168425-64-7

IUPAC Name: 2-(morpholin-4-yl)-4H-pyrimido[2,1-a]isoquinolin-4-one
Synonyms: 2-(morpholin-4-yl)pyrimido[2,1-a]isoquinolin-4-one
Molecular Weight (g/mol): 281.32
Molecular Formula: C16H15N3O2
InChi Key: BVRDQVRQVGRNHG-UHFFFAOYSA-N
SMILES: O=C1C=C(N=C2N1C=CC1=CC=CC=C21)N1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results