accessibility menu, dialog, popup

UserName

CAS RN 168127-49-9

NNN

CAS RN 168127-49-9

IUPAC Name: 9-[6-(9H-carbazol-9-yl)pyridin-2-yl]-9H-carbazole
Synonyms: 9-[6-(carbazol-9-yl)pyridin-2-yl]carbazole
Molecular Weight (g/mol): 409.49
Molecular Formula: C29H19N3
InChi Key: CUQGKGMUSQKHFO-UHFFFAOYSA-N
SMILES: C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC(=N1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results