missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(9H-carbazol-9-yl)pyridine 98.0+%, TCI America™

Catalog No. B4961200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4961200MG 200 mg
B49611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4961200MG Supplier TCI America Supplier No. B4961200MG
Only null left

Chemical Identifiers

CAS 168127-49-9
Molecular Formula C29H19N3
Molecular Weight (g/mol) 409.492
MDL Number MFCD28138085
InChI Key CUQGKGMUSQKHFO-UHFFFAOYSA-N
PubChem CID 23062165
IUPAC Name 9-(6-carbazol-9-ylpyridin-2-yl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=NC(=CC=C4)N5C6=CC=CC=C6C7=CC=CC=C75

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 200 mg
Formula Weight 409.49
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(9H-carbazol-9-yl)pyridine