accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 16732-09-5

H3COOBrBrBr

CAS RN 16732-09-5

IUPAC Name: 2,4,6-tribromophenyl hexanoate
Synonyms: 2,4,6-tribromophenyl hexanoate
Molecular Weight (g/mol): 428.95
Molecular Formula: C12H13Br3O2
InChi Key: WKADNUIXFNFRSW-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OC1=C(Br)C=C(Br)C=C1Br
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.