missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenyl Hexanoate 97.0+%, TCI America™

Catalog No. H011325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H011325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H011325G Supplier TCI America Supplier No. H011325G
Only null left

Chemical Identifiers

CAS 16732-09-5
Molecular Formula C12H13Br3O2
Molecular Weight (g/mol) 428.95
MDL Number MFCD00059464
InChI Key WKADNUIXFNFRSW-UHFFFAOYSA-N
Synonym Hexanoic Acid 2,4,6-Tribromophenyl Ester
PubChem CID 567165
IUPAC Name 2,4,6-tribromophenyl hexanoate
SMILES CCCCCC(=O)OC1=C(Br)C=C(Br)C=C1Br

Specifications

Color Yellow
Quantity 25 g
Formula Weight 428.95
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6-Tribromophenyl Hexanoate
EINECSNumber (9)-0289
TSCA No