accessibility menu, dialog, popup

UserName

H3CNHN

CAS RN 16731-68-3

IUPAC Name: 2-undecyl-1H-imidazole
Synonyms: 2-undecyl-1H-imidazole
Molecular Weight (g/mol): 222.38
Molecular Formula: C14H26N2
InChi Key: LLEASVZEQBICSN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1=NC=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results