accessibility menu, dialog, popup

UserName

CAS RN 16730-20-4

IUPAC Name: 5-nitro-1H-indole-2-carboxylic acid
Synonyms: 5-nitro-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H6N2O4
InChi Key: LHFOJSCXLFKDIR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results