accessibility menu, dialog, popup

UserName

HOOHN

CAS RN 1670-83-3

IUPAC Name: 1H-indole-7-carboxylic acid
Synonyms: 1H-indole-7-carboxylic acid
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: IPDOBVFESNNYEE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2NC=CC2=CC=C1
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)