accessibility menu, dialog, popup

UserName

CAS RN 165800-03-3

IUPAC Name: N-{[(5S)-3-[3-fluoro-4-(morpholin-4-yl)phenyl]-2-oxo-1,3-oxazolidin-5-yl]methyl}acetamide
Synonyms: linezolid zyvoxid linezlid zyvox
Molecular Weight (g/mol): 337.35
Molecular Formula: C16H20FN3O4
InChi Key: TYZROVQLWOKYKF-ZDUSSCGKSA-N
SMILES: CC(=O)NC[C@H]1CN(C(=O)O1)C1=CC=C(N2CCOCC2)C(F)=C1
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results