missing translation for 'onlineSavingsMsg'
Learn More

Linezolid, 98%

Catalog No. AC460592500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460592500 250 mg
AC460590010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460592500 Supplier Thermo Scientific Chemicals Supplier No. 460592500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Linezolid, Zyvox, CAS # 165800-03-3 is a synthetic oxazolidinone antibiotic. | CAS: 165800-03-3 | C16H20FN3O4 | 337.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Linezolid is an organoflourine compound and an acetamide

• Linezolid acts by inhibiting initiation complex formation by binding to the 50S ribosomal subunit

Applications

• Linezolid is effective against staphylococcal and enterococcal microbes

• It is most effective against gram-positive organisms, particularly methicillin, vancomycin and penicillin resistant ones

• Suitable for research on antibiotic resistance, cross-reactivity and drug mechanisms

Chemical Identifiers

CAS 165800-03-3
Molecular Formula C16H20FN3O4
Molecular Weight (g/mol) 337.35
InChI Key TYZROVQLWOKYKF-ZDUSSCGKSA-N
Synonym linezolid, zyvox, zyvoxid, s-n-3-3-fluoro-4-morpholinophenyl-2-oxooxazolidin-5-yl methyl acetamide, zyvoxam, linezolide, linezlid, zyvoxa, linezolid usan:inn, unii-isq9i6j12j
PubChem CID 441401
ChEBI CHEBI:63607
IUPAC Name N-[[(5S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide
SMILES CC(=O)NCC1CN(C(=O)O1)C2=CC(=C(C=C2)N3CCOCC3)F

Specifications

Melting Point 181.0°C to 183.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 337.35
Percent Purity 98%
Chemical Name or Material Linezolid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes damage to organs through prolonged or repeated exposure if inhaled.
Precautionary Statement GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Get medical attention/advice if you feel unwell.

RUO – Research Use Only