accessibility menu, dialog, popup

UserName

CAS RN 16545-54-3

IUPAC Name: tetradecyl 3-{[3-oxo-3-(tetradecyloxy)propyl]sulfanyl}propanoate
Synonyms: dimyristyl thiodipropionate
Molecular Weight (g/mol): 570.96
Molecular Formula: C34H66O4S
InChi Key: LVEOKSIILWWVEO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results