missing translation for 'onlineSavingsMsg'
Learn More

Ditetradecyl 3,3'-Thiodipropionate 94.0+%, TCI America™

Catalog No. T0911500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0911500G 500 g
T091125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0911500G Supplier TCI America Supplier No. T0911500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16545-54-3
Molecular Formula C34H66O4S
Molecular Weight (g/mol) 570.96
MDL Number MFCD00048462
InChI Key LVEOKSIILWWVEO-UHFFFAOYSA-N
PubChem CID 85489
IUPAC Name tetradecyl 3-{[3-oxo-3-(tetradecyloxy)propyl]sulfanyl}propanoate
SMILES CCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCC

Specifications

Melting Point 52°C
Color White
Quantity 500 g
Synonym Dimyristyl 3,3′-Thiodipropionate, 3,3′-Thiodipropionic Acid Dimyristyl Ester, 3,3′-Thiodipropionic Acid Ditetradecyl Ester
Formula Weight 570.96
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ditetradecyl 3,3′-Thiodipropionate
EINECSNumber (2)-0919&(2)-1399
TSCA Yes