accessibility menu, dialog, popup

UserName

Br

CAS RN 1647-26-3

IUPAC Name: (2-bromoethyl)cyclohexane
Synonyms: 2-bromoethylcyclohexane
Molecular Weight (g/mol): 191.11
Molecular Formula: C8H15Br
InChi Key: JRQAAYVLPPGEHT-UHFFFAOYSA-N
SMILES: BrCCC1CCCCC1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)