accessibility menu, dialog, popup

UserName

CAS RN 163927-30-8

IUPAC Name: 4-(3-isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benzoxadiazole
Synonyms: 4-(3-isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benzoxadiazole
Molecular Weight (g/mol): 291.29
Molecular Formula: C11H9N5O3S
InChi Key: CHKCHHTYBXOUKH-UHFFFAOYNA-N
SMILES: [O-][N+](=O)C1=CC=C(N2CCC(C2)N=C=S)C2=NON=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results