missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-NBD-Py-NCS 98.0+%, TCI America™

Catalog No. A5578100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5578100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5578100MG Supplier TCI America Supplier No. A5578100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[HPLC Labeling Reagent for e.e.Determination]

Chemical Identifiers

CAS 163927-30-8
Molecular Formula C11H9N5O3S
Molecular Weight (g/mol) 291.285
MDL Number MFCD02093435
InChI Key CHKCHHTYBXOUKH-ZETCQYMHSA-N
Synonym (S)-(+)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitrobenzofurazan, (S)-(+)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benzoxadiazole
PubChem CID 16218824
IUPAC Name 7-[(3S)-3-isothiocyanatopyrrolidin-1-yl]-4-nitro-2,1,3-benzoxadiazole
SMILES C1CN(CC1N=C=S)C2=CC=C(C3=NON=C23)[N+](=O)[O-]

Specifications

Quantity 100 mg
Formula Weight 291.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-NBD-Py-NCS [=(S)-(+)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benz
TSCA No
Recommended Storage Refrigerator