accessibility menu, dialog, popup

UserName

CAS RN 163451-81-8

IUPAC Name: 2-cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide
Synonyms: teriflunomide
Molecular Weight (g/mol): 270.21
Molecular Formula: C12H9F3N2O2
InChi Key: UTNUDOFZCWSZMS-UHFFFAOYSA-N
SMILES: CC(O)=C(C#N)C(=O)NC1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results