missing translation for 'onlineSavingsMsg'
Learn More

Teriflunomide 98.0+%, TCI America™

Catalog No. T32871G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32871G 1 g
T3287100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32871G Supplier TCI America Supplier No. T32871G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 163451-81-8
Molecular Formula C12H9F3N2O2
Molecular Weight (g/mol) 270.211
MDL Number MFCD00910058
InChI Key UTNUDOFZCWSZMS-YFHOEESVSA-N
Synonym N-[4-(Trifluoromethyl)phenyl]-2-cyano-3-hydroxycrotonamide, (Z)-2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-2-butenamide
PubChem CID 54684141
ChEBI CHEBI:68540
IUPAC Name (Z)-2-cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide
SMILES CC(=C(C#N)C(=O)NC1=CC=C(C=C1)C(F)(F)F)O

Specifications

Melting Point 229°C
Color White
Quantity 1 g
Formula Weight 270.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Teriflunomide
TSCA No
Recommended Storage Freezer (-20 °C)