accessibility menu, dialog, popup

UserName

CH3(S)O(S)(S)(R)HOHC

CAS RN 16320-04-0

IUPAC Name: (1R,3aS,3bS,11aS)-11a-ethyl-1-ethynyl-1-hydroxy-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: gestrinone gestrinonum dimetriose ethylnorgestrienone gestrinone (substance) gestrinone (product) aventis brand of gestrinone gestrinona tridomose florizel brand of gestrinone
Molecular Weight (g/mol): 308.42
Molecular Formula: C21H24O2
InChi Key: BJJXHLWLUDYTGC-ANULTFPQSA-N
SMILES: CC[C@]12C=CC3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@]2(O)C#C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results