missing translation for 'onlineSavingsMsg'
Learn More

Gestrinone 98.0+%, TCI America™

Catalog No. G0436100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0436100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G0436100MG Supplier TCI America Supplier No. G0436100MG
Only null left

Chemical Identifiers

CAS 16320-04-0
Molecular Formula C21H24O2
Molecular Weight (g/mol) 308.42
MDL Number MFCD00865561
InChI Key BJJXHLWLUDYTGC-ANULTFPQSA-N
PubChem CID 27812
IUPAC Name (1R,3aS,3bS,11aS)-11a-ethyl-1-ethynyl-1-hydroxy-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,11aH-cyclopenta[a]phenanthren-7-one
SMILES CC[C@]12C=CC3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@]2(O)C#C

Specifications

Melting Point 152°C
Color Yellow
Quantity 100 mg
Formula Weight 308.42
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Gestrinone
RTECSNumber JF7964000
TSCA No
Recommended Storage Refrigerator